Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054004.2 | NP_446456.1 | 4 | 813 | ||
GenBank mRNA | D87671.1 | - | 4 | 815 |