Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080896.1 | NP_543172.1 | 4 | 405 | ||
GenBank mRNA | AF367468.1 | - | 4 | 405 | ||
BC099792.1 | - | 484 | ||||
BC129087.1 | - | 623 | 8363754 |