Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085402.1 | NP_001078871.1 | 3 | 446 | ||
GenBank mRNA | EF100686.1 | - | 3 | 446 | ||
EU128746.1 | - | 3 | 452 | |||
FQ221935.1 | - | 537 |
Gene Symbol: Clec2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130402.2 | NP_569086.1 | 3 | 446 | ||
XM_008763309.1 | XP_008761531.1 | 4 | 943 | |||
GenBank mRNA | AF321552.1 | - | 5 | 632 | ||
EF100688.1 | - | 3 | 446 |
Gene Symbol: Clec2dl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085404.1 | NP_001078873.1 | 3 | 446 | ||
XM_006237453.2 | XP_006237515.1 | 4 | 479 | |||
XM_008763333.1 | XP_008761555.1 | 3 | 1505 | |||
GenBank mRNA | EF100689.1 | - | 3 | 446 | ||
FJ404471.1 | - | 3 | 446 |
Gene Symbol: Clec2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048075.1 | NP_001041540.1 | 3 | 350 | ||
GenBank mRNA | DQ168417.1 | - | 3 | 350 | ||
DQ168419.1 | - | 3 | 350 | |||
EF100690.1 | - | 3 | 350 | |||
EU128748.1 | - | 3 | 356 |
Gene Symbol: LOC689757 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101019.2 | NP_001094489.2 | 3 | 530 | ||
XM_008763332.1 | XP_008761554.1 | 3 | 833 | |||
GenBank mRNA | EF100687.1 | - | 3 | 500 | ||
FJ404472.1 | - | 3 | 530 |
Gene Symbol: LOC689770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142304.1 | NP_001135776.1 | 500 | |||
GenBank mRNA | EU128747.1 | - | 506 | |||
FJ404474.1 | - | 500 |