Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130748.1 | NP_570104.1 | 5, 6 | 535 | ||
XM_008765663.1 | XP_008763885.1 | 5, 6 | 530 | |||
GenBank mRNA | AF295535.1 | - | 5, 6 | 535 | ||
BC097292.1 | - | 6, 7 | 463 | 7462761 |