Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130814.1 | NP_570827.1 | 4, 5 | 612 | ||
GenBank mRNA | AB079374.1 | - | 4, 5 | 612 | ||
AF348446.1 | - | 3, 4 | 426 |