Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GALT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007255.2 | NP_009186.1 | 3 | 614 | ||
XM_006714816.3 | XP_006714879.1 | 3 | 546 | |||
XM_017008999.1 | XP_016864488.1 | 3 | 2737 | |||
GenBank mRNA | AB028600.1 | - | 3 | 561 | ||
AF142675.1 | - | 3 | 556 | |||
AJ005382.1 | - | 3 | 521 | |||
AK022566.1 | - | 1 | 1621 | |||
AK023506.1 | - | 3 | 557 | |||
AK095882.1 | - | 3 | 1221 | |||
AK123092.1 | - | 1 | 45 | |||
AY358578.1 | - | 3 | 597 | |||
BC007317.2 | - | 3 | 614 | 3049998 | ||
BC062983.1 | - | 3 | 581 | 4577572 | ||
BC072403.1 | - | 3 | 603 | 6094074 |