Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133416.1 | NP_596907.1 | 1 | 334 | ||
GenBank mRNA | AF378332.1 | - | 1 | 334 |