Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035237.1 | NP_001030314.1 | 10 | 937 | ||
NM_133524.2 | NP_598208.2 | 10 | 937 | |||
GenBank mRNA | AJ973227.1 | - | 10 | 937 | ||
BC078750.1 | - | 10 | 865 | 7123064 | ||
X54549.1 | - | 10 | 937 | |||
X62323.1 | - | 10 | 841 |