Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007260.2 | NP_009191.1 | 6, 7 | 411 | ||
XM_005245728.4 | XP_005245785.1 | 6, 7 | 883 | |||
XM_017000165.1 | XP_016855654.1 | 6, 7 | 778 | |||
GenBank mRNA | AF090423.1 | - | 6, 7 | 457 | ||
AF098668.1 | - | 6, 7 | 411 | |||
AK024724.1 | - | 2, 3 | 91 | |||
BC017034.2 | - | 6, 7 | 376 | 3845764 | ||
BC017193.2 | - | 6, 7 | 365 | 3138786 |
Gene Symbol: LYPLA2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033250.1 | 1 | 400 |