Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270862.1 | NP_001257791.1 | 4 | 218 | ||
NM_001270863.1 | NP_001257792.1 | 4 | 200 | |||
NM_001270864.1 | NP_001257793.1 | 3 | 158 | |||
NM_133583.2 | NP_598267.2 | 3 | 176 | |||
GenBank mRNA | AF334105.1 | - | 4 | 182 | ||
AF334106.1 | - | 3 | 140 | |||
AJ426424.1 | - | 4 | 231 | |||
AJ426425.1 | - | 3 | 189 | |||
AJ426426.1 | - | 4 | 194 | |||
AJ426427.1 | - | 3 | 152 | |||
FM045410.1 | - | 4 | 218 | |||
FM057759.1 | - | 3 | 158 |