Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270862.1 | NP_001257791.1 | 16 | 1745 | ||
NM_001270863.1 | NP_001257792.1 | 16 | 1727 | |||
NM_001270864.1 | NP_001257793.1 | 15 | 1685 | |||
NM_133583.2 | NP_598267.2 | 15 | 1703 | |||
GenBank mRNA | AF334105.1 | - | 16 | 1709 | ||
AF334106.1 | - | 15 | 1667 | |||
AJ426424.1 | - | 16 | 1758 | |||
AJ426425.1 | - | 15 | 1716 | |||
AJ426426.1 | - | 16 | 1721 | |||
AJ426427.1 | - | 15 | 1679 | |||
FQ212926.1 | - | 1 | 708 | |||
FQ213480.1 | - | 1 | 708 | |||
FQ216465.1 | - | 8 | 1093 | |||
FQ218864.1 | - | 1 | 708 |