Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007272.2 | NP_009203.2 | 4 | 307 | ||
XM_011540550.1 | XP_011538852.1 | 4 | 307 | |||
GenBank mRNA | AK289447.1 | - | 4 | 306 | ||
BC015118.1 | - | 4 | 313 | 3950170 | ||
BC059779.1 | - | 4 | 286 | |||
CR749296.1 | - | 2 | 117 | |||
S82198.1 | - | 4 | 284 | |||
Y13697.1 | - | 3 | 236 |