Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133617.2 | NP_598301.2 | 2 | 480 | ||
XM_008764736.1 | XP_008762958.1 | 2 | 498 | |||
GenBank mRNA | BC088113.1 | - | 2 | 480 | 7326099 | |
U55765.1 | - | 1 | 392 |
Gene Symbol: LOC100909524 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750227.3 | XP_003750275.1 | 2 | 498 | ||
XM_006240518.2 | XP_006240580.1 | 2 | 498 |