Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134361.1 | NP_599188.1 | 2 | 122 | ||
GenBank mRNA | U23377.1 | - | 2 | 122 |