Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Avpi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134373.2 | NP_599200.2 | 3 | 851 | ||
XM_006231383.2 | XP_006231445.1 | 3 | 917 | |||
GenBank mRNA | AJ294544.1 | - | 3 | 857 | ||
FQ220140.1 | - | 3 | 761 | |||
FQ220957.1 | - | 3 | 735 |