Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134374.2 | NP_599201.1 | 4 | 1350 | ||
GenBank mRNA | AB070897.1 | - | 4 | 1350 |