Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elovl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134382.1 | NP_599209.1 | 3, 4 | 332 | ||
XM_006243417.1 | XP_006243479.1 | 4, 5 | 497 | |||
GenBank mRNA | AB071985.1 | - | 3, 4 | 332 | ||
HQ404314.1 | - | 3, 4 | 332 |