Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem14c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135169.2 | NP_001128641.1 | 4, 5 | 321 | ||
NM_134395.2 | NP_599222.2 | 3, 4 | 534 | |||
XM_006253777.2 | XP_006253839.1 | 4, 5 | 524 | |||
GenBank mRNA | AF083396.1 | - | 3, 4 | 542 | ||
BC158854.1 | - | 3, 4 | 215 | 7936967 | ||
CB744788.1 | - | 4, 5 | 375 | |||
FQ211234.1 | - | 4, 5 | 311 | |||
FQ217516.1 | - | 4, 5 | 311 | |||
FQ218209.1 | - | 4, 5 | 317 | |||
FQ223586.1 | - | 4, 5 | 301 | |||
FQ223742.1 | - | 4, 5 | 311 | |||
FQ224495.1 | - | 4, 5 | 317 | |||
FQ225707.1 | - | 4, 5 | 291 | |||
FQ230083.1 | - | 4, 5 | 339 | |||
FQ230323.1 | - | 4, 5 | 320 | |||
FQ231364.1 | - | 4, 5 | 301 | |||
FQ233821.1 | - | 4, 5 | 309 |