Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Svop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134404.2 | NP_599231.2 | 7 | 763 | ||
XM_008769315.1 | XP_008767537.1 | 7 | 825 | |||
XM_008769316.1 | XP_008767538.1 | 7 | 1291 | |||
GenBank mRNA | AF060173.1 | - | 7 | 920 |