Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr7a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134407.1 | NP_599234.1 | 5, 6 | 786 | ||
GenBank mRNA | AB037424.1 | - | 5, 6 | 786 | ||
AF503514.1 | - | 5, 6 | 824 | |||
AJ271883.1 | - | 5, 6 | 789 | |||
BC061816.1 | - | 5, 6 | 789 | 5598724 | ||
FQ230711.1 | - | 2, 3 | 195 | |||
FQ231883.1 | - | 2, 3 | 195 |