Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100909664 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751234.3 | XP_003751282.2 | 6 | 1800 | ||
XM_006249680.2 | XP_006249742.1 | 6 | 1801 | |||
XM_006249681.2 | XP_006249743.1 | 6 | 1801 | |||
XM_006249682.2 | XP_006249744.1 | 6 | 1800 | |||
XM_006249683.2 | XP_006249745.1 | 6 | 1800 |
Gene Symbol: Cep170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769812.1 | XP_008768034.1 | 6 | 905 | ||
GenBank mRNA | AB038495.1 | - | 3 | 576 |