Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138522.1 | NP_612531.1 | 4 | 382 | ||
GenBank mRNA | D21095.1 | - | 4 | 382 |