Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138530.2 | NP_612539.2 | 182 | |||
XM_006256372.2 | XP_006256434.1 | 3, 4 | 497 | |||
GenBank mRNA | AY083160.1 | - | 194 | |||
BC078735.1 | - | 182 | 7126368 | |||
FQ209640.1 | - | 217 | ||||
FQ219037.1 | - | 217 |