Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnase2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138539.2 | NP_612548.1 | 5 | 825 | ||
GenBank mRNA | AF178975.1 | - | 5 | 826 | ||
BC091124.1 | - | 5 | 728 | 7375398 | ||
FQ216710.1 | - | 5 | 826 |