Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spdya | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138855.1 | NP_620210.1 | 6 | 924 | ||
XM_006239766.1 | XP_006239828.1 | 6 | 1158 | |||
XM_006239767.2 | XP_006239829.1 | 6 | 1122 | |||
XM_006239768.2 | XP_006239830.1 | 6 | 989 | |||
XM_008764502.1 | XP_008762724.1 | 6 | 1824 | |||
GenBank mRNA | AF492385.1 | - | 6 | 924 | ||
BC078729.1 | - | 5 | 847 | 7134092 |