Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138862.3 | NP_620217.2 | 5 | 418 | ||
XM_006240413.2 | XP_006240475.1 | 4 | 401 | |||
XM_006240414.2 | XP_006240476.1 | 5 | 493 | |||
XM_006240415.2 | XP_006240477.1 | 3 | 326 | |||
XM_008764737.1 | XP_008762959.1 | 4 | 372 | |||
GenBank mRNA | BC078728.1 | - | 5 | 408 | 7114267 | |
CK470849.1 | - | 1 | 82 | |||
CK600391.1 | - | 3 | 247 |