Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301203.1 | NP_001288132.1 | 2 | 377 | ||
NM_007364.3 | NP_031390.1 | 2 | 377 | |||
NR_125394.1 | 2 | 377 | ||||
XM_017022040.1 | XP_016877529.1 | 2 | 377 | |||
XM_017022041.1 | XP_016877530.1 | 2 | 326 | |||
GenBank mRNA | AJ132270.1 | - | 2 | 208 | ||
AK075529.1 | - | 2 | 280 | |||
AK222516.1 | - | 2 | 315 | |||
AK289434.1 | - | 2 | 296 | |||
AK297710.1 | - | 2 | 294 | |||
AK304148.1 | - | 2 | 283 | |||
AL109672.1 | - | 2 | 287 | |||
AY358974.1 | - | 2 | 287 | |||
BC000027.2 | - | 2 | 285 | 3507308 | ||
BC010853.1 | - | 2 | 322 | 3876465 | ||
BC017495.1 | - | 2 | 288 | 3921858 | ||
BC022232.1 | - | 2 | 312 | 3885305 | ||
BC111547.1 | - | 2 | 348 | 40005178 | ||
BM549126.1 | - | 2 | 214 | |||
BP260160.1 | - | 2 | 377 | |||
CR457384.1 | - | 2 | 189 |