Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033951.1 | NP_001029123.1 | 5 | 428 | ||
NM_138920.1 | NP_620275.1 | 5 | 428 | |||
XM_008764738.1 | XP_008762960.1 | 5 | 465 | |||
XM_008764739.1 | XP_008762961.1 | 5 | 465 | |||
GenBank mRNA | AB032932.1 | - | 5 | 428 | ||
AB097075.1 | - | 5 | 428 | |||
AB097076.1 | - | 5 | 428 | |||
AB097077.1 | - | 5 | 428 | |||
BC087712.1 | - | 5 | 434 | 7317608 |