Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007372.3 | NP_031398.2 | 5, 6 | 718 | ||
NM_203499.2 | NP_987095.1 | 4, 5 | 647 | |||
XM_006721657.1 | XP_006721720.1 | 5, 6 | 855 | |||
XM_017024111.1 | XP_016879600.1 | 6, 7 | 954 | |||
GenBank mRNA | AB036090.1 | - | 3, 4 | 443 | ||
AF083255.1 | - | 2, 3 | 103 | |||
AK095772.1 | - | 2, 3 | 105 | |||
AK292908.1 | - | 2, 3 | 230 | |||
AL527653.3 | - | 5, 6 | 712 | |||
AL545851.3 | - | 4, 5 | 615 | |||
BC015505.1 | - | 3, 4 | 428 | 3905607 | ||
BC078667.1 | - | 3, 4 | 386 | |||
BC093081.1 | - | 5, 6 | 682 | 30407786 |