Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nupl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139091.2 | NP_620791.1 | 2 | 300 | ||
XM_006252093.2 | XP_006252155.1 | 2 | 405 | |||
GenBank mRNA | AF000900.1 | - | 2 | 237 | ||
AF000901.1 | - | 1 | 193 | |||
BC085690.1 | - | 2 | 300 | 7190650 | ||
U63839.1 | - | 2 | 313 |