Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012064.3 | NP_036196.1 | 1 | 386 | ||
GenBank mRNA | BC074913.2 | - | 1 | 377 | 30915524 | |
BC117474.1 | - | 1 | 386 | 40126025 | ||
CD675949.1 | - | 1 | 382 | |||
KT966395.1 | - | 1 | 335 |