Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139106.1 | NP_620806.1 | 4 | 790 | ||
XM_006240845.2 | XP_006240907.1 | 5 | 1305 | |||
GenBank mRNA | BC161836.1 | - | 4 | 2450 | 9098303 | |
FQ214102.1 | - | 998 | ||||
FQ223899.1 | - | 4 | 838 | |||
U00926.1 | - | 4 | 790 |