Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139109.2 | NP_620809.1 | 2, 3 | 254 | ||
GenBank mRNA | AF277899.1 | - | 2, 3 | 257 | ||
BC081723.1 | - | 2, 3 | 248 | 7108549 |