Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPRL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077350.2 | NP_001070818.1 | 3, 4 | 320 | ||
NM_001243248.1 | NP_001230177.1 | 3, 4 | 595 | |||
NM_001243249.1 | NP_001230178.1 | 2, 3 | 627 | |||
GenBank mRNA | AK298795.1 | - | 2, 3 | 189 | ||
AK300736.1 | - | 3, 4 | 285 | |||
BC020392.1 | - | 2, 3 | 441 | |||
DA258664.1 | - | 3, 4 | 294 | |||
DA658076.1 | - | 3, 4 | 295 | |||
X90857.1 | - | 3, 4 | 481 |