Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc34a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139338.2 | NP_647554.1 | 3 | 238 | ||
XM_006233558.2 | XP_006233620.1 | 4 | 451 | |||
XM_006233559.2 | XP_006233621.1 | 4 | 567 | |||
XM_006233560.2 | XP_006233622.1 | 4 | 428 | |||
XM_006233561.2 | XP_006233623.1 | 3 | 341 | |||
XM_006233562.2 | XP_006233624.1 | 3 | 457 | |||
XM_006233564.2 | XP_006233626.1 | 3 | 321 | |||
XM_006233565.2 | XP_006233627.1 | 4 | 427 | |||
XM_006233566.2 | XP_006233628.1 | 3 | 433 | |||
XM_006233567.2 | XP_006233629.1 | 4 | 453 | |||
XM_008761577.1 | XP_008759799.1 | 4 | 516 | |||
GenBank mRNA | AB077042.1 | - | 3 | 243 |