Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144750.2 | NP_653351.1 | 4, 5 | 515 | ||
GenBank mRNA | AB009372.1 | - | 4, 5 | 516 | ||
BC098655.1 | - | 4, 5 | 485 | 7373402 |