Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Faim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144756.2 | NP_653357.1 | 12 | 1191 | ||
GenBank mRNA | AF044201.1 | - | 12 | 1140 | ||
BC087606.1 | - | 12 | 1191 | 7314068 |