Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gimap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033913.1 | NP_001029085.1 | 3 | 1091 | ||
NM_145680.3 | NP_663713.1 | 3 | 916 | |||
XM_006236457.2 | XP_006236519.1 | 3 | 1454 | |||
XM_006236458.2 | XP_006236520.1 | 3 | 1262 | |||
XM_008762871.1 | XP_008761093.1 | 4 | 1321 | |||
GenBank mRNA | AY055776.3 | - | 3 | 916 | ||
AY055777.2 | - | 1 | 803 | |||
AY550018.1 | - | 3 | 1091 | |||
AY550021.1 | - | 5 | 1609 | |||
AY550022.1 | - | 5 | 1434 | |||
AY550023.1 | - | 4 | 1326 | |||
BC092561.1 | - | 3 | 945 | 7130634 | ||
DQ125352.1 | - | 3 | 747 | |||
DQ125353.1 | - | 3 | 746 |