Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007204.4 | NP_009135.4 | 4 | 972 | ||
GenBank mRNA | AF106019.1 | - | 4 | 625 | ||
AF155803.1 | - | 4 | 620 | |||
AF171063.1 | - | 4 | 908 | |||
AK001506.1 | - | 4 | 634 | |||
AK301697.1 | - | 3 | 473 | |||
AK314312.1 | - | 4 | 617 | |||
AL133598.1 | - | 1 | 127 | |||
BC011556.1 | - | 4 | 625 | 4651697 | ||
BC031062.1 | - | 4 | 634 | 5273709 | ||
BC034953.2 | - | 4 | 943 | 4824324 |