Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110151.1 | NP_001103621.1 | 10, 11 | 1635 | ||
NM_152847.2 | NP_690060.2 | 10, 11 | 1635 | |||
GenBank mRNA | AB010245.1 | - | 10, 11 | 1635 | ||
AB051816.1 | - | 10, 11 | 1635 |