Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153294.1 | NP_695206.1 | 1 | 498 | ||
GenBank mRNA | AB084278.1 | - | 1 | 498 |