Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007214.4 | NP_009145.1 | 3 | 521 | ||
XM_011535399.2 | XP_011533701.1 | 3 | 270 | |||
XM_017010218.1 | XP_016865707.1 | 3 | 354 | |||
GenBank mRNA | AF100141.1 | - | 3 | 252 | ||
AJ011779.1 | - | 3 | 350 | |||
AK001465.1 | - | 3 | 384 | |||
AK074835.1 | - | 3 | 337 | |||
BC023598.2 | - | 3 | 432 | |||
BC047221.1 | - | 3 | 390 | 5277083 | ||
BC048287.1 | - | 3 | 377 | 5212007 | ||
BM764168.1 | - | 3 | 397 | |||
DB049187.1 | - | 3 | 543 | |||
KU178551.1 | - | 3 | 252 | |||
KU178552.1 | - | 3 | 252 |