Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007215.3 | NP_009146.2 | 4 | 943 | ||
XM_006721651.2 | XP_006721714.1 | 4 | 896 | |||
XM_017024072.1 | XP_016879561.1 | 4 | 896 | |||
GenBank mRNA | AF142992.1 | - | 4 | 926 | ||
AF177201.1 | - | 4 | 845 | |||
AF184344.1 | - | 4 | 926 | |||
AK307801.1 | - | 4 | 896 | |||
BC000913.2 | - | 4 | 691 | |||
BC009194.2 | - | 4 | 913 | 3949617 | ||
U94703.1 | - | 4 | 927 |