Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153311.1 | NP_695223.1 | 3 | 217 | ||
GenBank mRNA | AF537098.1 | - | 3 | 217 |