Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153317.1 | NP_695229.1 | 7 | 573 | ||
GenBank mRNA | BC158579.1 | - | 7 | 599 | 9091689 | |
U53475.1 | - | 7 | 573 |