Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007217.3 | NP_009148.2 | 5 | 575 | ||
NM_145859.1 | NP_665858.1 | 5 | 424 | |||
NM_145860.1 | NP_665859.1 | 4 | 330 | |||
XM_005247086.4 | XP_005247143.1 | 5 | 1310 | |||
XM_005247087.4 | XP_005247144.1 | 5 | 1315 | |||
XM_005247088.3 | XP_005247145.1 | 4 | 406 | |||
XM_006713485.3 | XP_006713548.1 | 5 | 470 | |||
XM_011512368.2 | XP_011510670.1 | 6 | 1383 | |||
XM_011512369.2 | XP_011510671.1 | 6 | 1388 | |||
XM_017005644.1 | XP_016861133.1 | 5 | 465 | |||
XM_017005645.1 | XP_016861134.1 | 5 | 1306 | |||
GenBank mRNA | AF022385.1 | - | 4 | 330 | ||
AK291130.1 | - | 4 | 361 | |||
BC002506.1 | - | 5 | 587 | 3050540 | ||
BC016353.1 | - | 5 | 428 | 4092244 | ||
CR457107.1 | - | 3 | 177 |