Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153622.2 | NP_705886.1 | 3 | 350 | ||
XM_006253941.2 | XP_006254003.1 | 6 | 1003 | |||
XM_006253942.2 | XP_006254004.1 | 6 | 1005 | |||
XM_006253944.2 | XP_006254006.1 | 6 | 992 | |||
XM_006253946.2 | XP_006254008.1 | 5 | 879 | |||
XM_006253947.2 | XP_006254009.1 | 6 | 1009 | |||
XM_006253948.2 | XP_006254010.1 | 1 | 880 | |||
XM_006253949.2 | XP_006254011.1 | 4 | 762 | |||
XM_006253950.2 | XP_006254012.1 | 6 | 1006 | |||
XM_006253951.2 | XP_006254013.1 | 5 | 880 | |||
XM_008771623.1 | XP_008769845.1 | 7 | 1080 | |||
XM_008771624.1 | XP_008769846.1 | 7 | 1068 | |||
XM_008771625.1 | XP_008769847.1 | 5 | 838 | |||
GenBank mRNA | AB025223.1 | - | 3 | 374 | ||
AB025224.1 | - | 3 | 374 | |||
BC078710.1 | - | 3 | 268 | 7110091 |