Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec11c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153628.2 | NP_705892.1 | 3 | 334 | ||
GenBank mRNA | AB022714.2 | - | 3 | 335 |