Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nalcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153630.1 | NP_705894.1 | 442 | |||
GenBank mRNA | AF078779.1 | - | 442 | |||
AY555273.1 | - | 460 | ||||
EF427668.1 | - | 3, 4 | 400 | |||
EF492985.1 | - | 3, 4 | 400 |