Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifi47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172019.2 | NP_742016.2 | 2 | 1343 | ||
XM_006246175.2 | XP_006246237.1 | 2 | 2375 | |||
XM_008767662.1 | XP_008765884.1 | 2 | 1563 | |||
GenBank mRNA | AF508023.1 | - | 2 | 1361 | ||
BC066660.1 | - | 2 | 1314 | 5623225 |